Learn More
NAALADase-like 2/NAALADL2 Mouse anti-Human, Alexa Fluor 700, Clone: 817225, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB4665N100UG
Catalog No. FAB4665N100
Add to cart
This item is not returnable. View return policy
Description
Description
NAALADase-like 2/NAALADL2 Monoclonal specifically detects NAALADase-like 2/NAALADL2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
NAALADase-like 2/NAALADL2
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q58DX5
NAALADL2
Mouse myeloma cell line NS0-derived recombinant human NAALADase-like 2/NAALADL2 Ser152-Asn795 Accession # Q58DX5
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human NAALADase-like 2/NAALADL2 in direct ELISAs.
Flow Cytometry
817225
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
inactive N-acetylated-alpha-linked acidic dipeptidase-like protein 2, NAALADase L2, NAALADase like 2, NAALADL2, N-acetylated alpha-linked acidic dipeptidase 2, N-acetylated alpha-linked acidic dipeptidase-like 2
Mouse
IgG1
RUO
Primary
254827
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)