Learn More
Spear Bio™ NAB-Sure™ SARS-CoV-2 Neutralizing Antibody Test Kit
This assay is an ultrasensitive, cell-free neutralizing antibody (NAb) test for immune response to SARS-CoV-2. The assay simplifies COVID-19 NAb testing from sampling through result, enabling the use of very small sample volumes using qPCR.
Manufacturer:  Spear Bio™ SPRNS01
Catalog No. 22-400-100
Add to cart
This item is not returnable. View return policy
Description
Description
NAB-Sure™ SARS-CoV-2 Assay is the first cell-free NAb assay with superior sensitivity and precision as compared to conventional cell-based approaches. It is a surrogate virus neutralization test (sVNT) which generates NAb (titer) measurements using a variety of real-time PCR systems.
  • Combines PRNT level accuracy with unparalleled sensitivity and lab-to lab consistency to assess plasma, serum and dried blood spots (DBS)
  • Tool for researchers and pharma developers working in vaccine efficacy testing, COVID-19 sero-surveillance and longitudinal antibody durability testing
Specifications
Specifications
Not Applicable
No
Yes
Non-sterile
SARS-CoV-2 Neutralizing Antibodies
Kit
RT-PCR