Learn More
NAB1 Mouse anti-Human, Clone: OTI1H3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF803233
Catalog No. 50-166-6388
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Acts as a transcriptional repressor for zinc finger transcription factors EGR1 and EGR2.
Specifications
Specifications
NAB1
Monoclonal
Unconjugated
NAB1
Lyophilized
NAB1
Human recombit protein fragment corresponding to amino acids 237-487 of human NAB1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1H3
PBS with 8% trehalose and no preservative; pH 7.3
Q13506
NGFI-A binding protein 1
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4664