Learn More
NAB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 185411AP
Catalog No. 50-173-2177
Add to cart
Description
Description
Acts as a transcriptional repressor for zinc finger transcription factors EGR1 and EGR2.
Specifications
Specifications
NAB1
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q13506, Q61122, Q62722
EGR 1 binding protein 1, NAB1, NGFI A binding protein 1
Rabbit
IgG
150 μL
-20°C
Primary
17936, 4664, 64824
Western Blot
Unconjugated
NAB1
Liquid
Nab1
NAB1 Fusion Protein Ag13407
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat