Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAC1 Mouse anti-Human, Clone: PCRP-NACC1-1A8, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen NAC1
Clone PCRP-NACC1-1A8
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Protein Array
Applications Flow Cytometry, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122489
View Documents
Novus Biologicals
NBP30750810UG
100 μg
Each for $352.89
Add to cart
 
NB122490
View Documents
Novus Biologicals
NBP30750820UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

NAC1 Monoclonal antibody specifically detects NAC1 in Human samples. It is validated for Flow Cytometry, Protein Array
Specifications

Specifications

NAC1
0.2 mg/ml
Flow Cytometry, Peptide Array
Purified
RUO
Human
BEND8, BTB (POZ) domain containing 14B, BTB/POZ domain-containing protein 14B, BTBD14B, NAC1BEN domain containing 8, NAC-1FLJ37383, nucleus accumbens associated 1, BEN and BTB (POZ) domain containing, nucleus accumbens-associated protein 1, transcriptional repressor NAC1
Recombinant full-length human NAC1 protein
Monoclonal
Protein A or G purified
PCRP-NACC1-1A8
Flow Cytometry 1-2 ug/million cells, Protein Array
Unconjugated
Mouse
Cancer, Chromatin Research, Stem Cell Markers, Transcription Factors and Regulators
10 mM PBS with 0.05% BSA
112939
IgG2a
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit