Learn More
BioVision IncSupplier Diversity Partner PicoProbe™ NADH Fluorometric Assay Kit: PicoProbe™ NADH Fluorometric Assay Kit: The most sensitive kit on the market. Fluorometric assay to measure NADH in animal tissues, cells & enzymatic reactions. 100 assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K338100
NAD and NADH exist in all living organisms and are important for energy transformation and maintenance of the redox state of cells or tissues. The reduced form, NADH is a driving force for cells and plays a critical role in cell regulation and repair processes. It is of increasing interest to be able to measure low level of NADH in samples or in enzymatic reactions. BioVision’s PicoProbe™ NADH Assay kit provides a highly sensitive method to detect low level of NADH (NAD is not detected in the reaction). There is no requirement to purify NADH from samples. The NADH Recycling Enzyme Mix specifically recognizes NADH (not NADPH) in the enzyme recycling reaction. The assay is rapid, simple and can measure less than 8 nM NADH in a variety of samples. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9703