Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NADKD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | NADKD1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP156734
|
Novus Biologicals
NBP156734 |
100 μL |
Each of 1 for $436.00
|
|
Description
NADKD1 Polyclonal specifically detects NADKD1 in Human samples. It is validated for Western Blot.Specifications
NADKD1 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
chromosome 5 open reading frame 33, FLJ30596, hypothetical protein LOC133686, MGC43298 | |
NADKD1 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q4G0N4 | |
133686 | |
Synthetic peptides corresponding to NADKD1 The peptide sequence was selected from the middle region of NADKD1. Peptide sequence RWLWRQRIRLYLEGTGINPVPVDLHEQQLSLNQHNRALNIERAHDERSEA. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title