Learn More
Selleck Chemical LLCSupplier Diversity Partner Nafamostat Mesylate 50mg 82956-11-4 FUT-175  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S138650MG
Nafamostat mesilate (FUT-175) is a synthetic serine protease inhibitor, used as an anticoagulant during hemodialysis. Nafamostat mesylate blocks activation of SARS-CoV-2 and is investigated as a new treatment option for COVID-19. Nafamostat Mesilate attenuates inflammation and apoptosis.
Catalog No. 50-628-7