Alfa Aesar™ Nafion™ D-520 dispersion, 5% w/w in water and 1-propanol, ≥1.00 meq/g exchange capacity

Preparation of thin films and coatings; proton exchange membranes

Manufacturer: Alfa Aesar™ 42118AE

Quantity 100mL

 View more versions of this product

Catalog No. AA42118AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 31175-20-9
    MDL Number MFCD00132790
    UN Number UN1987
    Synonym Perfluorosulfonic acid-PTFE copolymer, 5% w/w solution
    Chemical Name or Material Nafion™ D-520 dispersion, ≥0.92meq/g exchange capacity

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1987
    Health Hazard 2 GHS H Statement
    H318-H226-H336
    Causes serious eye damage.
    Flammable liquid and vapor.
    May cause drowsiness or dizziness.
    Quantity 100mL
    TSCA TSCA
    Physical Form Liquid
    MDL Number MFCD00132790
    Synonym Perfluorosulfonic acid-PTFE copolymer, 5% w/w solution
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
    Flash Point 24° (75°F)
    Density 0.93g/mL
    CAS 31175-20-9
    Color Colorless to yellow
    Concentration 5% w/w in water and 1-propanol
    Chemical Name or Material Nafion™ D-520 dispersion, ≥0.92meq/g exchange capacity

    GHS H Statement
    H318-H226-H336
    Causes serious eye damage.
    Flammable liquid and vapor.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit