Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nafion™ D-521 dispersion, 5% w/w in water and 1-propanol, ≥0.92 meq/g exchange capacity

Preparation of thin films and coatings; proton exchange membranes

Manufacturer:  Alfa Aesar™ 42117AE

Quantity 100mL

 View more versions of this product

Catalog No. AA42117AE


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

31175-20-9
UN1987
Nafion™ D-521 dispersion, ≥0.92meq/g exchange capacity
MFCD00132790
Perfluorosulfonic acid-PTFE copolymer, 5% w/w solution

Specifications

31175-20-9
Liquid
UN1987
Colorless to yellow
100mL
Nafion™ D-521 dispersion, ≥0.92meq/g exchange capacity
0.93g/mL
MFCD00132790
Perfluorosulfonic acid-PTFE copolymer, 5% w/w solution
5% w/w in water and 1-propanol
24° (75°F)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H318-H226-H336
Causes serious eye damage.
Flammable liquid and vapor.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit