Alfa Aesar™ Nafion™ D-521 dispersion, 5% w/w in water and 1-propanol, ≥0.92 meq/g exchange capacity

Preparation of thin films and coatings; proton exchange membranes

Manufacturer: Alfa Aesar™ 42117K4

Quantity 2.5L

 View more versions of this product

Catalog No. AA42117K4

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 31175-20-9
    MDL Number MFCD00132790
    UN Number UN1987
    Synonym Perfluorosulfonic acid-PTFE copolymer, 5% w/w solution
    Chemical Name or Material Nafion™ D-521 dispersion, ≥0.92meq/g exchange capacity

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Density 0.93g/mL
    Physical Form Liquid
    MDL Number MFCD00132790
    UN Number UN1987
    Synonym Perfluorosulfonic acid-PTFE copolymer, 5% w/w solution
    Color Colorless to yellow
    Concentration 5% w/w in water and 1-propanol
    Quantity 2.5L
    Flash Point 24° (75°F)
    TSCA TSCA
    CAS 31175-20-9
    Health Hazard 2 GHS H Statement
    H318-H226-H336
    Causes serious eye damage.
    Flammable liquid and vapor.
    May cause drowsiness or dizziness.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
    Chemical Name or Material Nafion™ D-521 dispersion, ≥0.92meq/g exchange capacity

    GHS H Statement
    H318-H226-H336
    Causes serious eye damage.
    Flammable liquid and vapor.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit