Learn More
Fuel Cell Earth Nafion Membrane NRE-212, 15cm x 15cm  DFS Item
Manufacturer:  Fuel Cell Earth NRE21215
Nafion Membrane NRE-212, 2 milsNafion Membrane is used to separate the anode and cathode compartment of Proton Exchange Membrane fuel cells and water electrolyzers. The thickness of this particular cation exchange membrane, Nafion 212 makes it suitable for many Hydrogen Air/Oxygen Fuel Cell applications. Nafion 212 is 50.8 micrometers (2 mil) thick. The membrane is positioned between a backing film and a coversheet.
Catalog No. 50-109-0472