Learn More
Nafion|r NR50, super acidic ion exchange resin catalyst, Thermo Scientific Chemicals
Description
Super acid catalyst Nafion™ NR50 is a perfluorinated ion-exchange membrane which has a wide variety of commercial uses. It is an acidic hydrogen ion form and superacid catalysts useful in a wide variety of synthetic applications. It finds application as a catalyst in alkylation and acylation reactions. It is also used in the production of sensors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Pellet |
Melting Point | 200°C |
Density | 2.1 g/mL |
Color | Brown to White |
Quantity | 50 g |
Solubility Information | Insoluble in water. |
Chemical Name or Material | Nafion™ NR50, Super acidic ion exchange resin catalyst |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.