Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nafion|r NR50, super acidic ion exchange resin catalyst, Thermo Scientific™

Click to view available options
Quantity:
10g
50g
31175-20-9
C9HF17O5S
544.135
MFCD00132790
FOYUGSIADQEOEK-UHFFFAOYSA-N
nafion, nafion nr 50 beads 10-35 mesh, perfluoro 2-2-sulfonylethoxy propyl vinyl ether-tetrafluoroethylene copolymer, tetrafluoroethylene-perfluoro 2-2-sulfonylethoxy propyl vinyl ether copolymer, 2-1-difluoro trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoroethanesulfonyl fluoride-tetrafl, ethanesulfonic acid, 1,1,2,2-tetrafluoro-2-1,2,2-trifluoro-1-trifluoromethyl-2-1,2,2-trifluorovinyl oxy ethoxy-, pol, ethanesulfonic acid, 2-1-difluoro trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoro-, polymer, nafion n-324 membrane, 0.15mm 0.006in thick, teflon fabric reinforced, nafion nre-212 membrane, 0.05mm thick, >=0.92 meq/g exchange capacity, nafion d-520 dispersion w/w in water and 1-propanol, >=1.00 meq/g exchange capacity
61889
1,1,2,2-tetrafluoroethene;1,1,2,2-tetrafluoro-2-[1,1,1,2,3,3-hexafluoro-3-(1,2,2-trifluoroethenoxy)propan-2-yl]oxyethanesulfonic acid
C(=C(F)F)(OC(C(C(F)(F)F)(OC(C(F)(F)S(=O)(=O)O)(F)F)F)(F)F)F.C(=C(F)F)(F)F
31175-20-9
C9HF17O5S
544.135
MFCD00132790
FOYUGSIADQEOEK-UHFFFAOYSA-N
nafion, nafion nr 50 beads 10-35 mesh, perfluoro 2-2-sulfonylethoxy propyl vinyl ether-tetrafluoroethylene copolymer, tetrafluoroethylene-perfluoro 2-2-sulfonylethoxy propyl vinyl ether copolymer, 2-1-difluoro trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoroethanesulfonyl fluoride-tetrafl, ethanesulfonic acid, 1,1,2,2-tetrafluoro-2-1,2,2-trifluoro-1-trifluoromethyl-2-1,2,2-trifluorovinyl oxy ethoxy-, pol, ethanesulfonic acid, 2-1-difluoro trifluoroethenyl oxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2-tetrafluoro-, polymer, nafion n-324 membrane, 0.15mm 0.006in thick, teflon fabric reinforced, nafion nre-212 membrane, 0.05mm thick, >=0.92 meq/g exchange capacity, nafion d-520 dispersion w/w in water and 1-propanol, >=1.00 meq/g exchange capacity
61889
1,1,2,2-tetrafluoroethene;1,1,2,2-tetrafluoro-2-[1,1,1,2,3,3-hexafluoro-3-(1,2,2-trifluoroethenoxy)propan-2-yl]oxyethanesulfonic acid
C(=C(F)F)(OC(C(C(F)(F)F)(OC(C(F)(F)S(=O)(=O)O)(F)F)F)(F)F)F.C(=C(F)F)(F)F

Super acid catalyst

Density 2.1 g/mL
Color Brown to White
Melting Point 200°C
Quantity 10g
Solubility Information Insoluble in water.
Sensitivity Hygroscopic
Physical Form Pellet
Chemical Name or Material Nafion™ NR50, Super acidic ion exchange resin catalyst

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures

RUO – Research Use Only