Alfa Aesar™ Nafion™ R-1100 resin, sulfonyl fluoride form

Thermoplastic-can be extruded or laminated

Manufacturer: Alfa Aesar™ 4462630

Quantity 250g

 View more versions of this product

Catalog No. AA4462630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 26654-97-7
    Molecular Formula Rf[OCF2CF(CF3)2]nOCF2CF2SO2F
    MDL Number MFCD05665455
    Synonym Perfluorosulfonyl fluoride-PTFE copolymer
    Chemical Name or Material Nafion™ R-1100 resin, Sulfonyl fluoride form

    Specifications

    Physical Form Pellet
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD05665455
    Health Hazard 1 Warning
    Quantity 250g
    TSCA TSCA
    Molecular Formula Rf[OCF2CF(CF3)2]nOCF2CF2SO2F
    CAS 26654-97-7
    Synonym Perfluorosulfonyl fluoride-PTFE copolymer
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point >200°C
    Chemical Name or Material Nafion™ R-1100 resin, Sulfonyl fluoride form

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit