Learn More
Selleck Chemical LLCSupplier Diversity Partner Naftopidil DiHCl 50mg 57149-08-3 KT-611  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S138750MG
Naftopidil DiHCl (KT-611) is a selective 5-HT1A and α1-adrenergic receptor antagonist with IC50 of 0.1 μM and 0.2 μM, respectively.
Catalog No. 50-628-9