Learn More
Selleck Chemical LLC
Naftopidil 50mg 57149-07-2 KT-611
Manufacturer: Selleck Chemical LLC S212650MG
Naftopidil (KT-611) is a selective α1-adrenergic receptor antagonist with Ki of 3.7 nM, 20 nM, and 1.2 nM for α1a, α1b, and α1d, respectively, used for the treatment of benign prostatic hyperplasia.