Learn More
Aviva Systems Biology Corp NAGS Antibody - C-terminal region (ARP51183_T100) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51183_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NAGS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NAGS. Alternate names: AGAS, ARGA, MGC133025ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60512