Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Nail Nipper, Sklar, 5-1/2 in., Sharp, Straight, Pliers with Lock, Stainless Steel. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21484
Econo Nail Nipper, Sklar, 5-1/2 in., Sharp, Straight, Pliers with Lock, Stainless Steel. The Econo Nail Nipper is used for removing portions of the toe nail or the nail plate. This product features double-spring, partially cross-serrated, plier handles with a locking mechanism that is released upon squeezing the handles. This lock is intended to keep the sharp blades closed when the instrument is not being used. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is 5-1/2 inches long and features angled, concave blades. It is sold in boxes of 12.
Catalog No. 50-143-7645