Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nalcn, Mouse anti-Rat, AP, Clone: S187-7, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB18378
Specifications
Nalcn | |
Monoclonal | |
Alkaline Phosphatase (AP) | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
Vgcnl1 | |
Mouse | |
100 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
S187-7 | |
Immunocytochemistry (1:100) Immunofluorescence (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
sodium leak channel, non-selective | |
Nalcn | |
Recombinant protein corresponding to amino acids 1659-1738 at C-terminus of rat Nalcn. | |
RUO | |
266760 | |
Store at -20°C. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction