Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalcn, Mouse anti-Rat, ATTO633, Clone: S187-7, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB18374

Catalog No. 89-338-299


Add to cart

Description

Description

Specifications

Specifications

Nalcn
S187-7
Immunocytochemistry (1:100) Immunofluorescence (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
sodium leak channel, non-selective
Vgcnl1
Mouse
IgG1
RUO
Antibody
Monoclonal
Rat
Immunocytochemistry, Immunofluorescence, Western Blot
ATTO633
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Liquid
Nalcn
Recombinant protein corresponding to amino acids 1659-1738 at C-terminus of rat Nalcn.
100 μg
Store at -20°C.
Primary
266760
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit