Learn More
Thermo Scientific™ Nalgene™ 180 Clear Plastic PVC Metric Tubing
Establish a leak-tight fit with metric fittings by using Nalgene 180 PVC Metric Tubing, manufactured from the highest-grade resins and special plasticizers.
Manufacturer:  Thermo Scientific™ 80010508
 View more versions of this product
Catalog No. 14-178-116
Add to cart
Description
Description
Establish a leak-tight fit with metric fittings by using by using Thermo Scientific™ Nalgene™ 180 Clear Plastic PVC Metric Tubing. Manufactured from the highest-grade resins and special plasticizers. Available in true metric sizes, labelled and marked for easy cutting except where noted.
Features:
  • Lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins and plasticizers
  • Resists hardening and discoloration and a wide range of chemicals—demonstrates dimensional stability
  • Excellent for a wide variety of small benchtop applications, this tubing is transparent, nontoxic and non-contaminating
  • Marked with I.D. and regulatory compliance as well as at regular intervals for easy identification and measurement (Cat. No. 8001-0102, -0204, -0305, and -0406 are not printed)
Ordering Information:
In packs of 25m (10m for largest three sizes)
Specifications
Specifications
Food-grade material complies with USP VI and USDA regulations.
37 psig at 125°F (52°C), 74 psig at 73°F (23°C)
0.19 in.
PVC
8 mm
25 m
1.5 mm
25
Clear
Autoclavable
5 mm
0.31 in.
82 ft.
0.05 in.
Flexible Tubing
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. Warning: Not intended for medical use. Nalgene 180 and 980 PVC tubing contains DEHP [Bis (2-ethylhexyl)] phthalate, a commonly used plasticizer. It is known to the state of California Envirnmental Agency that this chemical causes cancer or reproductive toxicity.