Learn More
Thermo Scientific™ Nalgene™ 180 Clear Plastic PVC Metric Tubing Available on GSA/VA Contract for Federal Government customers only.
Establish a leak-tight fit with metric fittings by using Nalgene 180 PVC Metric Tubing, manufactured from the highest-grade resins and special plasticizers.
$135.20 - $715.50
Specifications
Certifications/Compliance Food-grade material complies with USP VI and USDA regulations.
Overall Length (English) 82 ft.
View More Specs
Products 8
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Maximum Pressure Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Maximum Pressure Price Quantity & Availability  
14-176-228 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
80010305
3 mm 5 mm 80 psig at 73°F (23°C) Case for $135.20
Add to cart
14-176-229
View Documents
Thermo Scientific™
80010406
4 mm 6 mm 66 psig at 73°F (23°C) Case of 25 for $190.20
Add to cart
14-178-116
View Documents
Thermo Scientific™
80010508
5 mm 8 mm 37 psig at 125°F (52°C), 74 psig at 73°F (23°C) Case for $302.50
Add to cart
14-176-238
View Documents
Thermo Scientific™
80011014
10 mm 14 mm 56 psig at 73°F (23°C) Case for $630.50
Add to cart
14-178-117
View Documents
Thermo Scientific™
80011216
12 mm 16 mm 50 psig at 73°F (23°C) Case for $715.50
Add to cart
14-178-115 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
80011520
15 mm 20 mm 50 psig at 73°F (23°C) Case for $311.50
Add to cart
14-176-236 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
80010812
8 mm 12 mm 66 psig at 73°F (23°C) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
14-176-233
View Documents
Thermo Scientific™
80010610
6 mm 10 mm 80 psig at 73°F (23°C) This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Establish a leak-tight fit with metric fittings by using by using Thermo Scientific™ Nalgene™ 180 Clear Plastic PVC Metric Tubing. Manufactured from the highest-grade resins and special plasticizers. Available in true metric sizes, labelled and marked for easy cutting except where noted.
Features:
  • Lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins and plasticizers
  • Resists hardening and discoloration and a wide range of chemicals—demonstrates dimensional stability
  • Excellent for a wide variety of small benchtop applications, this tubing is transparent, nontoxic and non-contaminating
  • Marked with I.D. and regulatory compliance as well as at regular intervals for easy identification and measurement (Cat. No. 8001-0102, -0204, -0305, and -0406 are not printed)
Ordering Information:
In packs of 25m (10m for largest three sizes)
Warranty: 90 days. For warranty terms, please contact technicalsupport@thermofisher.com.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Food-grade material complies with USP VI and USDA regulations.
82 ft.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. Warning: Not intended for medical use. Nalgene 180 and 980 PVC tubing contains DEHP [Bis (2-ethylhexyl)] phthalate, a commonly used plasticizer. It is known to the state of California Envirnmental Agency that this chemical causes cancer or reproductive toxicity.