Learn More
Thermo Scientific™ Nalgene™ 180 Clear Plastic PVC Vacuum Tubing Available on GSA/VA Contract for Federal Government customers only.
Make practical and efficient use of this Nalgene 180 vacuum tubing, thick-walled PVC which can withstand a full vacuum (80mm Hg) at room temperature.
Manufacturer:  Thermo Scientific™ 80000055
 View more versions of this product
Catalog No. 14-176-20
  Check Availability
Add to cart
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. Note:
Not intended for medical use. Nalgene 180 and 980 PVC tubing contain DEHP [Bis (2-ethylhexyl)] phthalate, a commonly used plasticizer. It is known to the state of California Environmental Agency that this chemical causes cancer or reproductive toxicity.
Specifications
Description
Make practical and efficient use of Thermo Scientific™ Nalgene™ 180 Clear Plastic Vacuum Tubing, thick-walled PVC which can withstand a full vacuum (80mm Hg) at room temperature. Material is Food-Grade and complies with USP VI and USDA regulations.
  • Withstands a full vacuum (80mm Hg)
  • Autoclavable/Transparent
Description & Specifications
Specifications
PVC
3/16 in.
50 ft.
15.24m
31psig
Material is Food-Grade and complies with USP VI and USDA regulations.
50
3/16 in.
9/16 in.
3/16 in.
0.48cm
180 PVC Vacuum Tubing 3/16 x 9/16 x 3/16
62psig
50
3/16 in.