Learn More
Thermo Scientific™ Nalgene™ 689 Polypropylene Tubing Available on GSA/VA Contract for Federal Government customers only.
Perform a variety of applications with the excellent chemical resistance to several acids and alkalis of Thermo Scientific? Nalgene? 689 Polypropylene Tubing.
Manufacturer:  Thermo Scientific™ 80200250
 View more versions of this product
Catalog No. 14-176-159
  Check Availability
Add to cart
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Perform a variety of applications with the excellent chemical resistance to several acids and alkalis of translucent and semirigid Thermo Scientific™ Nalgene™ 689 Polypropylene Tubing. This polypropylene tubing with durometer of 75 (Shore D) is unaffected by most solvents at ambient temperatures. Food-grade materials comply with 21 CFR 177.1520 for food packaging.
  • Unaffected by most solvents at ambient temperatures
  • Should not be exposed to halogenated hydrocarbons for an extended period
  • Semirigid Durometer 75 (shore D)
  • Autoclavable
Ordering Information:
In packs of 100ft. (30.5m)
Description & Specifications
Specifications
Polypropylene
3/16 in.
100 ft.
30.48m
55psig
Food-Grade materials comply with 21 CFR 177.1520 for food packaging
100
3/16 in.
1/4 in.
1/32 in.
0.08cm
689 Polypropylene Tubing 3/16 x 1/4 x 1/32
110psig
100
1/32 in.