Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Autoclavable Plastic Desiccators Products for Science Education

With polypropylene body and clear polycarbonate cover.

$425.00

Specifications

Color Blue with Clear
Autoclavable Yes
Material Polypropylene
For Use With (Application) High resistance to all solid desiccants
Resistance To all solid desiccants
View More Specs

Alert:

Do not use concentrated acids as desiccants.


Products 1
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Height (English) Exterior Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Height (English) Exterior Price Quantity & Availability  
S521043 Products for Science Education
View Documents
Thermo Scientific™
53090250/EMD
9.68 in. 13 in. 10.32 in.
Case of 4 for $425.00
Add to cart
 
Description

Description

  • Virtually unbreakable cover has a recessed rim to keep grease off of benchtop when set down
  • 140mm model comes with aluminum plate
  • 230mm model requires plate (not supplied)

Specifications

Specifications

Blue with Clear
Polypropylene
To all solid desiccants
Yes
High resistance to all solid desiccants
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit