Learn More
Thermo Scientific™ Nalgene™ Polypropylene, Closure with Barbed Bulkhead Fittings, 1/2 inch
Catalog Number 0292314W
Click to view available options
0.5 in. (13mm)
For Use With: Nalgene culture vessels
Use Thermo Scientific™ Nalgene™ Closures with Barbed Bulkhead Fittings to customize most Nalgene bottle and carboy closures for aseptic transfer of liquids.
  • Size 38mm-430 closures are predrilled allowing for easy, safe in-house assembly
  • Barbed at both ends to accept tubing inside and outside the container
  • Molded of white, autoclavable polypropylene, ready for in-house sterilization
Includes:
Instructions, drilling template and a set of two panel-mounted barbed fittings, acetal nuts and silicone gaskets.
Ordering Information:
Barbed bulkhead fittings also sold separately (6149-0001 and 6149-0002).
TRUSTED_SUSTAINABILITY
Product Type Closure
Closure Size 38-430mm
Description Closure with Barbed Bulkhead Fitting
Diameter (English) Inner Tubing 1/2 in.
Material Polypropylene
No. per Case 4
No. per Pack 4
For Use With (Equipment) Nalgene culture vessels