Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles
GSA_VAGREENER_CHOICE

Catalog No. 0340917C Shop All Thermo Scientific Products
Change view
Click to view available options
Closure Color:
Blue
Green
Red
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color
03-409-17C Red
03-409-17F Yellow
03-409-17E Blue
03-409-17D Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03-409-17C Supplier Thermo Scientific™ Supplier No. 24225500
  • $168.50
    $134.65 / Pack of 6

    Save $33.85

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $421.00
    $336.65 / Case of 4 PK

    Save $84.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Simplify content identification with these narrow-mouth wash bottles that feature colored closures with an integrated stem.

High quality squeeze bottles with color-coded closures for easy identification. Thermo Scientific™ Nalgene™ Color-Coded LDPE Wash Bottles are made from lab quality low density polyethylene (LDPE) for dependably low leachables and chemical compatability with a wide range of lab reagents. Enjoy smooth, precision, sputter-free fluid dispensing with an ergonomic soft squeeze. Ideal for rinsing tubes and lab apparatus, bringing volumetric flasks to final volume and other precision rinsing and solution dispensing needs. The dispensing nozzle is molded in one piece with the bottle closure eliminating risk of separation when the bottle is squeezed. Cut back tip to increase flow.

  • Color-coded top allows you to coordinate reagents with industry storage codes.
  • Tip can be cut back to provide increased flow.
  • Tight fit of draw tube and closure prevents separation when squeezing bottle.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 500 mL
Color Natural
Description With stem and draw tube
Material LDPE
No. per Case 24
No. per Pack 6
Closure Color Red
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 224 mm
Height (English) 8.8 in.
Closure Material Polypropylene
Closure Size 28 mm
Diameter (Metric) 72.9 mm
Closure Type Screw Cap
Through the Closure Dispensing Yes
Autoclavable Not Autoclavable
Capacity (English) 16 oz.
Shape Cylindrical
Product Type Color-coded Wash Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.