Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Fluorinated HDPE, Carboy with Closure

Click to view available options
Capacity (Metric):
10 L
20 L

For Use With: Enhance long-term container performance, prevent permeation loss and yield lower extractables ; Closure Material: HDPE; Color: White (Translucent)

Enhance long-term container performance, prevent permeation loss and yield lower extractables with Thermo Scientific™ Nalgene™ Fluorinated Carboys with Closure, for use with most acids, alkalis and aggressive organic solvents. These safe, durable and cost-efficient carboys featuring fluorinated surfaces (inside and outside), for improved barrier properties and reduced solvent absorption and penetration. Fluorination enhances long-term container performance and prevents or reduces permeation loss.

  • Leakproof†
  • Heavy-duty walls are safe, durable and resistant to splitting and punctures
  • HDPE screw caps included

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

No. per Pack 1
Autoclavable Not Autoclavable
Color White (Translucent)
Mouth Style Narrow
Type Carboy with Handle
Closure Material HDPE
No. per Case 6
For Use With (Application) Enhance long-term container performance, prevent permeation loss and yield lower extractables
Material Fluorinated HDPE
Capacity (English) 2.6 gal.
Height (Metric) With Closure 389 mm
Diameter (Metric) Inner Neck 64 mm
Capacity (Metric) 10 L
Closure Size 83 B
Show More Show Less