Learn More
Thermo Scientific™ Nalgene™ Glass Prefilters Available on GSA/VA Contract for Federal Government customers only.
Increase filtration speed and throughput of highly viscous or heavily particulated solutions when using theses prefilters with Nalgene MF75 filter units or bottle top filters.
Manufacturer:  Thermo Scientific™ DS02817500
 View more versions of this product
Catalog No. 09-740-24
Add to cart
Description
Description
Increase filtration speed and throughput of highly viscous or heavily particulated solutions by using Thermo Scientific™ Nalgene™ Glass Fiber Prefilter Discs in conjunction with any Nalgene™ MF75 filter units or bottle top filters.
  • Glass fiber prefilter discs
  • For Nalgene 50, 75 and 90mm dia. reusable filtration products Cat. Nos. 300, DS0310, DS0330, DS0315, DS0320
Ordering Information:
The 75mm size is included in cases of Nalgene 500 and 1000mL nylon membrane and surfactant-free cellulose acetate (SFCA) membrane filter units.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Glass-fiber
Prefilters
100
75mm
75mm
100
Glass
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.