Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Thermo Scientific™ Nalgene™ Large Cylindrical HDPE Containers with Covers Available on GSA/VA Contract for Federal Government customers only.
Store and transport dry and solid materials, specimens and general lab supplies. These stackable containers feature positive-lock lids that form a tight seal.
Manufacturer:  Thermo Scientific™ 71420030
 View more versions of this product
Catalog No. 11-843K
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Store and transport dry and solid materials, specimens and general lab supplies using Thermo Scientific™ Nalgene™ Large Cylindrical HDPE Containers with Covers. Containers are stackable and have positive-lock lids that seal tight for storing both dry and solid materials.
Features:
  • High-density polyethylene containers provide excellent chemical and temperature resistance
  • Easy-grip molded-in handles are comfortable and durable for lifting and moving
  • Positive-lock lid provides a tight seal for transport or storage
  • NSF approved for food and beverage storage
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
8 gal.
Container Large Round w/Cover HDPE 30 L
39 cm
30.5 cm
1
HDPE
NSF approved for food and beverage storage.
15.5 in.
12 in.
6
30 L
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.