Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Capacity (Metric):
2 L
4 L
8 L
Closure Size:
38-430 mm
53 B

Store corrosive chemicals and distilled water with Thermo Scientific™ Nalgene™ Large Narrow-Mouth PPCO Bottles, which are resistant to most acids, bases and alcohols.

Store corrosive chemicals and distilled water with Thermo Scientific™ Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure, which are resistant to most acids, bases and alcohols. Large sizes of these polypropylene copolymer bottles have a built-in shoulder loop.

Features:
  • 4 and 8L feature a built-in shoulder loop on larger sizes for attaching identification tag
  • Lighter weight than glass alternatives make for easy lifting
  • Translucence allows for easy viewing of liquid levels
  • Linerless caps provide the ultimate in leakproof protection without the use of liner that can wrinkle, cause leaks, or contaiminate your reagents
  • Autoclavable/Leakproof†

Includes:

Polypropylene screw caps

Note:

Completely disengage threads or remove closure before autoclaving. Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Height (Metric) 249 mm
Closure Size 38-430 mm
Shape Round
Closure Type Screw Cap
Mouth Narrow
Closure Liner Linerless
Closure Material Polypropylene
Diameter (Metric) 121 mm
Capacity 0.5 gal., 2 L
Capacity (Metric) 2 L
Capacity (English) 0.5 gal.
Color Natural Translucent
Description Large Narrow-Mouth Bottle PP 2L
Diameter (Metric) Inner Neck 25 mm
Material PPCO
No. per Case 6
No. per Pack 1
Sterility Non-sterile
Closure Color White
Neck Finish 38 mm
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.