Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Thermo Scientific™ Nalgene™ Large Polypropylene Waste Containers with Cover Available on GSA/VA Contract for Federal Government customers only.
Hold and sterilize refuse, clothing, equipment, labware and contaminated material with these large polypropylene waste containers featuring secure, friction-fit closures.
$693.50
Specifications
Certifications/Compliance Meet OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogensAutoclave upright with lid on.
Autoclavable Autoclavable
Material Polypropylene
Quantity 1/Pk., 4/Cs.
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Price Quantity & Availability  
15-353-6 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
69200120
45 L 36 cm Each for $693.50
Case of 4 EA for $1,727.00
N/A
15-353-5
View Documents
Thermo Scientific™
69200060
23 L 28 cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Hold and sterilize refuse, clothing, equipment, labware and contaminated material with Thermo Scientific™ Nalgene™ Large Polypropylene Waste Containers with Cover. These containers feature a secure, friction-fit closure and molded-in handgrips. They meet OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens.
  • Molded-in handgrips make lifting easy
  • Autoclavable/Biohazard
Note:
Autoclave this container upright with the lid on.
Warranty: 90 days. For warranty terms, please contact technicalsupport@thermofisher.com.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Meet OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogensAutoclave upright with lid on.
Autoclavable
Polypropylene
1/Pk., 4/Cs.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.