Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nalgene™ Narrow-Mouth Opaque Amber HDPE Bottles with Closure: Shrink-Wrapped Trays Available on GSA/VA Contract for Federal Government customers only.

Experience easy handling with Nalgene Opaque Amber HDPE Bottles, conveniently packaged in shrink-wrap trays ready to feed into filling lines for various reagents.

Manufacturer:  Thermo Scientific™ 3220850001

 View more versions of this product

Catalog No. 02-923-85


  Check Availability
Add to cart

Disclaimers:

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


Specifications

Description

Experience easy handling with Thermo Scientific™ Nalgene Opaque Amber HDPE Bottles, conveniently packaged in shrink-wrap trays ready to feed into filling lines for various reagents. Bottles include polypropylene closures.

  • Opaque amber container provides protection for light-sensitive reagents
  • Bottles are packed in rigid trays, allowing for easy handling
  • Opaque amber HDPE with opaque amber polypropylene closures
  • Leakproof†

Ordering Information:

Bottles are packed in shrink-wrapped trays with closures packed in separate polybags within the same carton. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Description & Specifications

Specifications

1 oz., 30mL
1 oz.
Polypropylene (PP)
Narrow-Mouth Packaging Bottles
HDPE
70
20-415mm
61mm
20mm
Screw closure (bulk packed, unassembled)
30mL
Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Product meets the requirements for light-resistant containers per USP latest edition
Amber Opaque
14mm
1050
Opaque amber
34mm
Narrow
Round
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit