Learn More
Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes Available on GSA/VA Contract for Federal Government customers only.
Conduct high-speed centrifugation of aggressive chemicals with these tubes, suited for chloroform and phenol extractions in refrigerated machines up to 50,000 x g.
$144.60 - $233.00
Specifications
Bottom Shape Round
Closure Material Tefzel ETFE
Autoclavable Yes
Graduations None
Material PTFE FEP
View More Specs
Alert:
Note: Must be filled to 100% of total capacity for centrifugation
Products 3
Catalog Number Mfr. No. Diameter (Metric) Outside Capacity (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Outside Capacity (Metric) Length (Metric) Price Quantity & Availability  
05-562-16C Available on GSA/VA Contract for Federal Government customers only. View Documents Thermo Scientific™
31140010
Pack of 2 for $144.60
Case of 5 PK for $452.00
Add to cart
05-562-16A Available on GSA/VA Contract for Federal Government customers only. Accessories View Documents Thermo Scientific™
31140030
25.7mm 30mL Pack of 2 for $209.00
Case of 5 PK for $658.50
Add to cart
05-562-16B Available on GSA/VA Contract for Federal Government customers only. Accessories View Documents Thermo Scientific™
31140050
28.8mm 50mL Pack of 2 for $233.00
Case of 5 PK for $727.00
Add to cart
Description
Description
Conduct high-speed centrifugation of aggressive chemicals with Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes, specifically suited for chloroform and phenol extractions in refrigerated machines up to 50,000 x g.
Features:
  • Teflon™ FEP, Tefzel™ ETFE screw closure
  • Withstand temperatures from -100° to +150°C (-148° to +302°F)
  • Leakproof† Tefzel™ ETFE closures prevent material loss under ordinary use for most spins <10,000 x g
  • Sealing caps available for 30mL and 50mL size to assure leakproof service especially at high speeds >10,000 x g or when spinning hazardous materials
  • Autoclavable/Biohazard/Leakproof†/PTFE FEP
Ordering Information:
Optional sealing caps available — DS3131-0020 (for 30mL tube) and DS3131-0024 (for 50mL tube). Cat. No. 3114-0050 has a brim capacity of only 42mL but is designed to fit standard rotor.
Includes:
Tefzel ETFE screw caps. Additional sealing cap assembly available separately (DS3131-series).
Warranty: 90 days
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Round
Yes
PTFE FEP
Non-sterile
No
Tefzel ETFE
None
50,000 x g
10
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.