Learn More
Thermo Scientific™ Nalgene™ PMP Economy Plastic Graduated Cylinders Available on GSA/VA Contract for Federal Government customers only.
Easily dispense liquids with the generous pour spouts on these PMP economy graduated cylinders. These clear cylinders exhibit good chemical and impact resistance.
$11.68 - $49.45
Specifications
Material PMP
No. per Pack 1
View More Specs
Products 7
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Graduations Price Quantity & Availability  
08-572-7A Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
36650010
10 mL 0.2 mL Each for $11.68
Case of 24 EA for $175.50
Add to cart
08-572-7B Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
36650025
25 mL 0.5 mL Each for $14.23
Case of 18 EA for $159.50
Add to cart
08-572-7C Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
36650050
50 mL 1 mL Each for $17.68
Case of 18 EA for $199.20
Add to cart
08-572-7D Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
36650100
100 mL 1 mL Each for $21.35
Case of 12 EA for $159.50
Add to cart
08-572-7E Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
36650250
250 mL 2 mL Each for $28.00
Case of 12 EA for $210.00
Add to cart
08-572-7F Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
36650500
500 mL 5 mL Each for $39.80
Case of 8 EA for $198.50
Add to cart
08-572-7G Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
36651000
1000 mL 10 mL Each for $49.45
Case of 6 EA for $185.10
Add to cart
Description
Description
Easily dispense liquids with the generous pour spouts on Thermo Scientific™ Nalgene™ PMP Economy Graduated Cylinders. These clear polymethylpentene cylinders exhibit good chemical and impact resistance and include easy to read molded-in graduations that won't rub off.
Features:
  • Transparent, break-resistant and non-wetting
  • Economical and recyclable—recommended for educational labs
  • No meniscus to confuse readings allow for accurate measurements
  • Good temperature resistance up to 135°C (275°F)
  • Large, ribbed, rounded base provides reinforcement and stability to keep from rolling or tipping
Warranty: 90 days. For warranty terms, please contact technicalsupport@thermofisher.com.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
PMP
1
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.