Learn More
Thermo Scientific™ Nalgene™ Powder Funnels
Won't chip, crack or break.
$49.25 - $92.00
Catalog Number | Mfr. No. | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
S34682
|
Thermo Scientific™
42520150/EMD |
716 mL | 13.6 cm |
Pack of 4 for $49.25
Case of 6 PK for $270.00
|
|
||||
S34680
|
Thermo Scientific™
42520080/EMD |
115 mL | 8.5 cm |
Pack of 12 for $92.00
Case of 3 PK for $250.00
|
|
||||
S34679
|
Thermo Scientific™
42520065/EMD |
68 mL | 6.7 cm |
Pack of 12 for $78.00
Case of 3 PK for $214.00
|
|
||||
S34681
|
Thermo Scientific™
42520100/EMD |
243 mL | 10.6 cm | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Tough, autoclavable polypropylene funnels
- Transfer liquids or powders
- External ribs allow for easy pouring
Specifications
Polypropylene | |
Yes |