Thermo Scientific™ Nalgene™ PPCO Amber High Profile Closures with Color Coders for Micro Packaging Vials: Sterile, Bulk Pack

Quickly identify samples using these high profile closures. Offered sterile in a variety of colors, these closures have no O-ring to fall out or contaminate contents.

Manufacturer: Thermo Scientific™ 3428250114

 View more versions of this product

Catalog No. 03-390-5

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.


    Specifications

    Type Closure
    Product Line Nalgene™
    Material Polypropylene Copolymer,Polypropylene copolymer (PPCO)
    Color Green,Green
    Closure Size 11mm
    No. per Case 1000
    No. per Pack 500
    Description High-profile closures w/color coders for micro packaging vials
    Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Micro Packaging Vials and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI. Amber Micro Packaging Vials meet the requirements for light-resistant containers per USP current edition when used with amber vials.
    Neck Finish 11mm
    Pre-sterilized Yes
    Item Description Green coder, 11mm, bulk pack

    Quickly identify samples using Thermo Scientific™ Nalgene™ PPCO Amber High Profile Closures with Color Coders for Micro Packaging Vials. Offered in a variety of colors, closures have no O-ring to fall out or contaminate contents.

    Features:

    • Vials (listed separately) and polypropylene copolymer closures are pressure-tested together at 7.5psig for air shipment.
    • Closures are for use with amber PPCO micro packaging vials to provide barrier for light-sensitive reagents.
    • Permanently attached, color-coded inserts are free of toxic heavy metals.
    • Inserts do not come in contact with vial contents.
    • Threaded, screw closure are sterile to 10-6 SAL, RNase/DNase Free, Non-cytotoxic, non-pyrogenic and leakproof† when used with Nalgene Micro Packaging Vials.

    Ordering Information:

    Micro Packaging Vial Closures are bulk packed without vials, 500 per inner bag, 1,000 per case. Order sterile amber vials separately (Cat. No. 342805).

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit