Learn More
Thermo Scientific™ Nalgene™ Autoclavable PPCO Wash Bottles
Description
Reduce the risk of contamination when dispensing sterile water or reagents by using autoclavable Thermo Scientific™ Nalgene™ PPCO Wash Bottles. Made from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. These squeeze bottles are flexible, contact-clear, and designed to dispense through the top with a closure and stem molded in one piece for reliable service. Bent stem dispensing tube retains its bend after autoclaving but bend angle can be modified by hand while warm if desired. The tight fit of the draw tube and closure prevents separation when squeezing bottle.
- Polypropylene screw/stem and draw tube
- Tip can be cut back to provide increased flow
Specifications
Diameter (Metric) | 72.6 mm |
Closure Type | Cap |
Closure Size | 28 mm |
Closure Material | Polypropylene |
Through the Closure Dispensing | Yes |
Height (Metric) | 213 mm |
Height (English) | 8.4 in. |
Product Type | Wash Bottle |
Capacity | 16 oz., 500 mL |
Capacity (Metric) | 500 mL |
Show More |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.