Learn More
Thermo Scientific Nalgene Rapid-Flow Sterile Single Use Vacuum Filter Units
Increase filtration efficiency. PES membranes and exclusive Rapid-Flow design provide fast flow rates and high throughput while defending against cell culture contamination.
Manufacturer: Thermo Scientific 1630020
Description
Increase filtration efficiency. Thermo Scientific; Nalgene Nalgene; Nalgene Rapid-Flow; Nalgene disposable Filter Units feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid Flow Filter Units and Bottle Tops are the best choice for the filtration of cell culture media, serum, additives and buffers.
Specifications
Yes | |
Filter Units | |
Nylon | |
12 | |
0.2μm | |
500mL |
Sterile | |
90mm | |
Nylon | |
12 | |
500mL |
Promotions
-
Thermo Scientific
Spend More, Get More on Labware
Buy up to $2,500 in Nalgene or Nunc Labware, get up to $1,200 more to spend at no additional cost.: 06-30-2021
-
Thermo Scientific
Get 1 Extra Case of Nalgene Filters When You Buy 3
Buy three cases of Thermo Scientific Nalgene Rapid Flow Filters, get a fourth at no additional cost.: 06-30-2021
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.