Learn More
Thermo Scientific™ Nalgene™ Sealing Caps for Oak Ridge Centrifuge Tubes

Use these sealing caps to ensure leakproof service of centrifuge bottles at their recommended maximum rated speeds.
$43.80 - $69.20
Specifications
Description | Sealing Caps for Oak Ridge Centrifuge Tubes |
---|---|
No. per Case | 2 |
No. per Pack | 2 |
Catalog Number | Mfr. No. | Closure Size | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Closure Size | For Use With (Equipment) | Price | Quantity & Availability | ||||
05-563-2B
|
Thermo Scientific™
DS31320020 |
20 mm | 28 and 30 mL Tubes |
Case of 2 for $43.80
|
|
05-563-2C
![]() |
Thermo Scientific™
DS31320024 |
24 mm | 50 mL Tubes |
Case of 2 for $55.60
|
|
05-563-2D
|
Thermo Scientific™
DS31320058 |
58 mm | 250 mL Tubes |
Case of 2 for $64.90
|
|
05-563-2E
|
Thermo Scientific™
DS31320063 |
63 mm | 500 mL and 1 L Tubes |
Case of 2 for $69.20
|
![]() |
Description
Ensure leakproof† service of Thermo Scientific™ Nalgene™ centrifuge bottles at their recommended maximum rated speeds by using Nalgene Sealing Caps for Oak Ridge Centrifuge Tubes. They are also recommended for use when spinning hazardous materials.
Compatibility:- 20 mm size – for use with Nalgene Oakridge tubes, 30ml volume
- 24 mm size – for use with Nalgene Oakridge tubes, 35ml and 50ml volumes
- 58 mm size - For use with Nalgene Centrifuge Bottles 3140-0250 and 3141-0250
- 63 mm size - For use with Nalgene Centrifuge Bottles 3140-0500, 3141-0500, 3120-1000, 3120-1010, 3122-1000 and 3122-1010
Note:
Before autoclaving, set closure on top of the container without engaging the threads.
Specifications
Sealing Caps for Oak Ridge Centrifuge Tubes | |
2 | |
2 |
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.