Learn More
Thermo Scientific Nalgene Suspended Magnetic Stir Bars
Use Nalgene Floating Magnetic Stir Bars for constant low-speed stirring action in tissue culture applications.
Manufacturer: Thermo Scientific DS66304000
Description
Constantly stir tissue culture applications at a low speed with Thermo Scientific Nalgene Suspended Magnetic Stir Bars.
Features:- Legs can be compressed to fit through 1.5in. (38mm) openings
- Stir bar is slightly raised off the bottom surface of the vessel to minimize grinding effect on cells
- Consistant action at low speeds
- Excellent resistance to chemicals and temperatures up to 150°C
- Alnico V magnet encapsulated in Teflon TFE with Tefzel ETFE bracket
Note:
Disassemble for cleaning.
Specifications
Suspended Magnetic Stir Bar; Stirbar Floating TFE/ETFE 155 (dia.) x 31mm | |
4000mL beaker | |
31mm | |
Magnetic |
6.125 in. | |
1.25 in. | |
2/Cs.; 1/Pk. | |
155mm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.