Thermo Scientific™ Nalgene™ Teflon™ FEP Wash Bottles Available on GSA/VA Contract for Federal Government customers only.

Ensure contamination-free dispensing with chemical-resistant Thermo Scientific™ Nalgene™ Teflon™ FEP Solvent Wash Bottles, molded entirely of fluoropolymers.

Manufacturer: Thermo Scientific™ 24030500

 View more versions of this product

Catalog No. 03-409-12E

  Check Availability
Add to Cart

Alert

† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Item Description Wash Bottle Teflon® FEP 500mL
    Capacity (Metric) 500mL
    Capacity (English) 16 oz.,16 oz.
    Closure Material ETFE
    Closure Size 38-430mm,38-430mm
    Closure Type Screw
    Material FEP
    No. per Case 4
    No. per Pack 1
    Capacity 16 oz., 500mL

    Ensure contamination-free dispensing with Thermo Scientific™ Nalgene™ Teflon™ FEP Solvent Wash Bottles, useful for trace element work. These phthalate-free bottles offer excellent chemical resistance and are molded entirely of fluoropolymers. Designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service. The precision-molded tip provides a straight, steady stream and ultimate control without sputtering.

    • With Tefzel™ ETFE screw closure/stem and draw tube
    • Demonstrates excellent chemical resistance to nearly any chemical in your lab including solvents—won't become brittle or crack
    • Inert fluoropolymer construction is low in extractables and low binding—useful for trace element work
    • Tip can be cut back to provide increased flow
    • Tight fit of draw tube and closure prevents separation when squeezing bottle
    • Autoclavable/Leakproof†/Transparent

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit