Learn More
Thermo Scientific™ Nalgene™ Torque Wrench Fittings for HDPE Closures Available on GSA/VA Contract for Federal Government customers only.
Assure accurate torque readings when applying closures onto bottles with Nalgene Torque Wrench Fittings for HDPE Closures.
Manufacturer:  Thermo Scientific™ 21951053
 View more versions of this product
Catalog No. 14-178-125
  Check Availability
Add to cart
Disclaimers:
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Assure accurate torque readings when applying closures onto bottles with Thermo Scientific™ Nalgene Torque Wrench Fittings for HDPE Closures. Proper torquing of Nalgene closures is critical to leakproof† sealing.
  • Epoxy
  • Specifically designed for Nalgene HDPE closures
Compatible with:
Thermo Scientific Nalgene Torque Wrenches (2195-6169 and -6177) † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Description & Specifications
Specifications
1/4 in.
Drive: 1/4 in.; 53-415
1
Nalgene™ Torque Wrenches,Nalgene Torque Wrenches (Cat. Nos. 2195-6169 and -6177)
Fittings
Nalgene Torque Wrenches (Cat. Nos. 2195-6169 and -6177)
53-415mm,53-415mm
1
Epoxy