Learn More
Thermo Scientific™ Nalgene™ Torque Wrench Fittings for HDPE Closures
Choose these torque wrench fittings designed for specific HDPE closures to achieve accurate torque readings when applying closures onto bottles.
Supplier:  Thermo Scientific™ 21951183
 View more versions of this product
Catalog No. 14-178-122
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Choose Thermo Scientific™ Nalgene™ Torque Wrench Fittings for HDPE Closures to achieve accurate torque readings when applying closures onto bottles. Proper torquing of Nalgene closures is critical to leakproof† sealing.
  • Epoxy
  • Specifically designed for Nalgene HDPE closures
Compatible with:
Thermo Scientific Nalgene Torque Wrenches (2195-6169 and -6177) † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fittings
3/8 in.
Epoxy
83B
Nalgene Torque Wrenches (Cat. Nos. 2195-6169 and -6177)
Product Suggestions
Product Suggestions