Learn More
Thermo Scientific™ Nalgene™ Torque Wrench Fittings for HDPE Closures
Assure accurate torque readings when applying closures onto bottles with Nalgene Torque Wrench Fittings for HDPE Closures.
$60.97 - $398.50
Specifications
For Use With (Equipment) Nalgene Torque Wrenches (Cat. Nos. 2195-6169 and -6177)
Material Epoxy
Type Fittings
View More Specs
Products 9
Catalog Number Mfr. No. Closure Size Type Price Quantity & Availability  
Catalog Number Mfr. No. Closure Size Type Price Quantity & Availability  
14-178-129
View Documents
Thermo Scientific™
21951020
20-415mm Fittings
Each for $98.60
Add to cart
02-925-110
View Documents
Thermo Scientific™
21951021
20-415mm Fittings
Case for $60.97
Add to cart
03-313-67
View Documents
Thermo Scientific™
21951024
24-415mm Fittings
Each for $100.30
Add to cart
14-178-130
View Documents
Thermo Scientific™
21951028
28-415mm Fittings
Case for $95.20
Add to cart
14-178-127
View Documents
Thermo Scientific™
21951038
38-415mm Fittings
Each for $125.50
Add to cart
14-178-128
View Documents
Thermo Scientific™
21951043
43-415mm Fittings
Case for $116.70
Add to cart
14-178-125
View Documents
Thermo Scientific™
21951053
53-415mm Fittings
Each for $126.70
Add to cart
14-178-121
View Documents
Thermo Scientific™
21951153
53B Fittings
Each for $237.50
Add to cart
14-178-122
View Documents
Thermo Scientific™
21951183
83B Fittings
Case for $398.50
Add to cart
Description
Description
Assure accurate torque readings when applying closures onto bottles with Thermo Scientific™ Nalgene Torque Wrench Fittings for HDPE Closures. Proper torquing of Nalgene closures is critical to leakproof† sealing.
  • Epoxy
  • Specifically designed for Nalgene HDPE closures
Compatible with:
Thermo Scientific Nalgene Torque Wrenches (2195-6169 and -6177)
Warranty: 90 days
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Specifications
Nalgene Torque Wrenches (Cat. Nos. 2195-6169 and -6177)
Epoxy
Fittings
SDS
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.