Learn More
Thermo Scientific™ Nalgene™ Torque Wrench Fittings for Polypropylene closure
Assure accurate torque readings when applying closures onto bottles with Nalgene Torque Wrench Fittings for Polypropylene Closures.
$99.90 - $456.00
Specifications
Material Epoxy
Type Fittings
View More Specs
Products 11
Catalog Number Mfr. No. Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Closure Size Price Quantity & Availability  
14-178-104
View Documents
Thermo Scientific™
21950020
20-415mm Case for $99.90
Add to cart
03-313-54
View Documents
Thermo Scientific™
21950024
24-415mm Each for $101.80
Add to cart
03-313-55
View Documents
Thermo Scientific™
21950028
28-415mm Each for $102.30
Add to cart
14-178-107
View Documents
Thermo Scientific™
21950038
38-415mm Case for $187.50
Add to cart
14-178-106
View Documents
Thermo Scientific™
21950043
43-415mm Each for $191.80
Add to cart
14-178-109
View Documents
Thermo Scientific™
21950048
48-415mm Each for $191.80
Add to cart
14-178-108
View Documents
Thermo Scientific™
21950053
53-415mm Each for $198.70
Add to cart
14-178-110
View Documents
Thermo Scientific™
21950063
63-415mm Case for $149.00
Add to cart
14-178-112
View Documents
Thermo Scientific™
21950153
53B Case for $252.00
Add to cart
14-178-113
View Documents
Thermo Scientific™
21950183
83B Each for $456.00
Add to cart
14-178-132
View Documents
Thermo Scientific™
21950438
38-430mm Case for $190.60
Add to cart
Description
Description
Assure accurate torque readings when applying closures onto bottles with Thermo Scientific™ Nalgene Torque Wrench Fittings for Polypropylene Closures. Proper torquing of Nalgene closures is critical to leakproof† sealing.
  • Epoxy
  • Specifically designed for Nalgene polypropylene closures
Compatible with:
Thermo Scientific Nalgene Torque Wrenches (2195-6169 and -6177)
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Warranty: 90 days
Specifications
Specifications
Epoxy
Fittings
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.