Thermo Scientific™ Nalgene™ Torque Wrench Fittings for Teflon™ closures
Assure accurate torque readings when applying closures onto bottles with Nalgene Torque Wrench Fittings for Teflon Closures.
Manufacturer: Thermo Scientific™ 21952038
 View more versions of this product
Catalog No. 14-178-154
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With (Equipment) Nalgene™ Torque Wrenches
    Type Fittings
    Description Torque wrench fittings for Tefzel™ closures
    Drive Size 1/4 in.
    No. per Case 1
    No. per Pack 1
    Closure Size 38-415mm
    For Use With Nalgene Torque Wrenches (Cat. No. 2195-6169)
    Material Epoxy
    Item Description Torque Fitting, Closure 38-415, ETFE
    Assure accurate torque readings when applying closures onto bottles with Thermo Scientific™ Nalgene Torque Wrench Fittings for Teflon Closures. Proper application torque is essential for leakproof† performance.
    • Durable epoxy resin construction
    • Specifically designed for Nalgene ETFE closures
    Compatible with:
    Compatible with Thermo Scientific Nalgene torque wrench 2195-6169 † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
  • Description & Specifications
Related Products
Product Content Feedback