Learn More
Thermo Scientific™ Nalgene™ Torque Wrench Fittings for ETFE Closures
Assure accurate torque readings when applying closures onto bottles with Nalgene™ Torque Wrench Fittings for ETFE closures.
$94.90 - $193.40
Specifications
For Use With (Equipment) Nalgene Torque Wrenches (Cat. No. 2195-6169)
Material Epoxy
No. per Case 1
No. per Pack 1
Type Fittings
View More Specs
Products 9
Catalog Number Mfr. No. Closure Size Price Quantity & Availability  
Catalog Number Mfr. No. Closure Size Price Quantity & Availability  
14-178-150
View Documents
Thermo Scientific™
21952020
20-415mm
Each for $94.90
Add to cart
14-178-151
View Documents
Thermo Scientific™
21952024
24-415mm
Case for $94.90
Add to cart
14-178-152
View Documents
Thermo Scientific™
21952028
28-415mm
Case for $111.70
Add to cart
14-178-153
View Documents
Thermo Scientific™
21952033
33-415mm
Each for $101.24
Add to cart
14-178-154
View Documents
Thermo Scientific™
21952038
38-415mm
Case for $188.30
Add to cart
14-178-155
View Documents
Thermo Scientific™
21952043
43-415mm
Each for $182.80
Add to cart
14-178-156
View Documents
Thermo Scientific™
21952048
48-415mm
Case for $188.40
Add to cart
14-178-157
View Documents
Thermo Scientific™
21952053
53-415mm
Case for $193.40
Add to cart
14-178-158
View Documents
Thermo Scientific™
21952438
38-430mm
Each for $188.30
Add to cart
Description
Description
Assure accurate torque readings when applying closures onto bottles with Thermo Scientific™ Nalgene™ Torque Wrench Fittings for ETFE Closures. Proper application torque is essential for leakproof† performance.
  • Durable epoxy resin construction
  • Specifically designed for Nalgene ETFE closures
Compatible with:
Compatible with Thermo Scientific Nalgene torque wrench 2195-6169 † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Warranty: 90 days
Specifications
Specifications
Nalgene Torque Wrenches (Cat. No. 2195-6169)
1
Fittings
Epoxy
1
1/4 in.
SDS
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.