Thermo Scientific™ Nalgene™ Torque Wrenches Available on GSA/VA Contract for Federal Government customers only.
Assure accurate torque readings when applying closures with Nalgene Torque Wrench Fittings.
Manufacturer: Thermo Scientific™ 21956169
 View more versions of this product
Catalog No. 03-313-50
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Type Torque Wrench
    Torque 0 to 75 lb.
    Description Torque wrench
    Drive Size 1/4 in.
    For Use With Nalgene Torque Wrench Fittings (Series 2195)
    No. per Case 1
    No. per Pack 1
    Item Description Drive : 1/4 in.; Torque: 0 to 75 lbs.
    Assure accurate torque readings when applying closures with Thermo Scientific™ Nalgene Torque Wrench Fittings. Proper torquing of Nalgene closures is critical to leakproof† sealing.
    These wrenches can be used to confirm capping machine settings, on-line inspection and to check torques. Must be used with Nalgene torque wrench fittings.
    Ordering Information:
    Order fittings that match closure resin.
    Compatible with:
    Thermo Scientific Nalgene Torque Wrench Fittings (Series 2195) † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
  • Description & Specifications
Related Products
Product Content Feedback